Learn More
SHP2 Mouse, Unlabeled, Clone: 79, BD
Mouse Monoclonal Antibody
£188.32 - £289.26
Specifications
| Antigen | SHP2 |
|---|---|
| Clone | 79 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|
15835549
|
BD Biosciences
610621 |
50 μg |
£196.00 £188.32 / 1 set Save £7.68 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15845549
|
BD Biosciences
610622 |
150 μg |
£510.00 £289.26 / 1 set Save £220.74 43% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SHP2 (also known as PTP1D, Syp, SHPTP2 and PTP2C) is a member of the cytosolic class of protein-tyrosine phosphatases (PTPs). SHP2 has been reported to contain two SH2 domains where both SH2 domains are N-terminal to the PTP catalytic domain. The expression of SHP2 has been reported to be highest in brain, heart, and kidney. The PTPs are thought to function with other protein-tyrosine kinases to maintain intracellular protein phosphotyrosine homeostasis and cell cycle progression. The presence ofSH2 domains in SHP2 (PTP1D) has prompted speculation that binding to specific phosphorylated tyrosine residues is key to its function. SHP2 (PTP1D) is tyrosine- phosphorylated and activated in response to stimulation with EGF or PDGF.
Spécification
| SHP2 | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 | |
| Affinity Purified | |
| PTP1D, Syp, SHPTP2 and PTP2C |
| 79 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Canine, Chicken, Frog, Human, Mouse, Rat | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.