missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SHOX Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA537220

Product Code. 15584301

  • £447.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody detects endogenous protein at a molecular weight of 29 kDa. Purity is >95% by SDS-PAGE.

Homeodomain proteins (HP) are transcriptional regulators that coordinate the expression of genes involved in development, differentiation and cellular transformation. HPs are characterized by a conserved domain of 60 amino acid residues that recognize and bind a site in the regulatory region of the target gene. SHOX, a member of the bicoid subfamily of the paired homeobox family, controls fundamental aspects of growth and development and undergoes splicing resulting in two isoforms, SHOXA and SHOXB. SHOXA is expressed in skeletal muscle, placental, pancreas, heart and bone marrow fibroblast and, to a lesser extent, in kidney and lung. SHOXB is highly expressed in osteogenic cells, but not expressed in brain, kidney, liver or lung. Defects in the gene SHOXX cause Leri-Weill dyschondrosteosis (LWD) and Langer mesomelic dysplasia (LMD).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SHOX
Polyclonal
Unconjugated
SHOX
GCFX; growth control factor, X-linked; PHOG; Pseudoautosomal homeobox-containing osteogenic protein; short stature homeobox; short stature homeobox protein; Short stature homeobox-containing protein; SHOX; SHOXY; SS
Rabbit
Antigen affinity chromatography
RUO
6473
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
O15266
SHOX
A synthetic peptide corresponding to residues in human SHOX.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.