Learn More
SHIP1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA532582
Description
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is tonsil tissue.
SHIP-1 p145 (SH2-containing-inositol phosphatase), an effector molecule, associates with activated Fms oncogenes. SHIP-1 contains two phosphotyrosine-binding domains (PTB), a unique amino terminal SH2 domain, a proline-rich region, and two highly conserved motifs found among inositol phosphate 5-phosphatases. SHIP-1 displays both phosphatidylinositol 3,4,5-triphosphate and inositol 1, 3, 4, 5 -tetrakisphosphate polyphosphate 5-phosphatase activity. Evidence suggests that SHIP-1 may modulate Ras signaling in addition to inositol signaling pathways.
Specifications
| SHIP1 | |
| Polyclonal | |
| INPP5D | |
| SHIP, SHIP1, SIP-145, hp51CN, SH2 containing inositol phosphatase, SH2 domain-containing inositol 5′-phosphatase 1, SH2 domain-containing inositol phosphatase 1, SH2 domain-containing inositol-5′-phosphatase 1, SHIP-1, inositol polyphosphate-5-phosphatase of 145 kDa, p150Ship, phosphatidylin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3635 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Q92835 | |
| INPP5D | |
| Synthetic peptide corresponding to C-terminus of human SHIP-1. | |
| 500 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.