Learn More
sex hormone-binding globulin, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SHBG.
Brand: Abnova H00006462-A01.50uL
Description
This gene encodes a steroid binding protein that was first described as a plasma protein secreted by the liver but is now thought to participate in the regulation of steroid responses. The encoded protein binds each steroid molecule as a dimer formed from identical or nearly identical monomers. The use of alternate promoters and alternatively spliced transcripts have been described. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: DPPAVHLSNGPGQEPIAVMTFDLTKITKTSSSFEVRTWDPEGVIFYGDTNPKDDWFMLGLRDGRPEIQLHNHWAQLTVGAGPRLDDGRWHQVEVKMEGDSSpecifications
| sex hormone-binding globulin | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant SHBG. | |
| SHBG | |
| ABP/MGC126834/MGC138391/TEBG | |
| Mouse | |
| 50 μL | |
| Yes | |
| 6462 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_001040 | |
| SHBG | |
| SHBG (NP_001031, 41 a.a. ∼ 140 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.