missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH3-domain GRB2-like 1 (A02), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SH3GL1.
Brand: Abnova H00006455-A02.50uL
This item is not returnable.
View return policy
Description
Sequence: KASQLVSEKVGGAEGTKLDDDFKEMEKKVDVTSKAVTEVLARTIEYLQPNPASRAKLTMLNTVSKIRGQVKNPGYPQSEGLLGECMIRHGKELGGESNFGDALLDAGESpecifications
| SH3-domain GRB2-like 1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant SH3GL1. | |
| SH3GL1 | |
| CNSA1/EEN/MGC111371/SH3D2B/SH3P8 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 6455 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_003025 | |
| SH3GL1 | |
| SH3GL1 (NP_003016, 12 a.a. ∼ 119 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction