Learn More
SH3-domain binding protein 4, Mouse, Clone: 2B6, Abnova™
Mouse monoclonal antibody raised against a partial recombinant SH3BP4.
Brand: Abnova H00023677-M11.100ug
Description
This gene encodes a protein with 3 Asn-Pro-Phe (NPF) motifs, an SH3 domain, a PXXP motif, a bipartite nuclear targeting signal, and a tyrosine phosphorylation site. This protein is involved in cargo-specific control of clathrin-mediated endocytosis, specifically controlling the internalization of a specific protein receptor. [provided by RefSeq
Sequence: MAAQRIRAANSNGLPRCKSEGTLIDLSEGFSETSFNDIKVPSPSALLVDNPTPFGNAKEVIAIKDYCPTNFTTLKFSKGDHLYVLDTSGGEWWYAHNTTEMGYIPSSYVQSpecifications
| SH3-domain binding protein 4 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC057396 | |
| SH3BP4 | |
| SH3BP4 (AAH57396, 1 a.a. ∼ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Cell Cycle | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 2B6 | |
| Mouse monoclonal antibody raised against a partial recombinant SH3BP4. | |
| SH3BP4 | |
| BOG25/TTP | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 23677 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.