missing translation for 'onlineSavingsMsg'
Learn More

SH2 domain containing 3C, Mouse, Clone: 3C5, Abnova™

Mouse monoclonal antibody raised against a partial recombinant SH2D3C.

Brand:  Abnova H00010044-M07.100ug

Product Code. 16101716

  • £303.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence: MTAVGRRCPALGSRGAAGEPEAGSDYVKFSKEKYILDSSPEKLHKELEEELKLSSTDLRSHAWYHGRIPREVSETLVQRNGDFLIRDSLTSLGDYVLTCRWRNQALHFKI
Specifications

Specifications

SH2 domain containing 3C
Monoclonal
Unconjugated
PBS with no preservative; pH 7.4
NM_005489
SH2D3C
SH2D3C (NP_005480, 1 a.a. ∼ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Signal Transduction
Primary
Human
Antibody
ELISA, Western Blot
3C5
Mouse monoclonal antibody raised against a partial recombinant SH2D3C.
SH2D3C
CHAT/FLJ39664/NSP3/PRO34088
Mouse
Affinity Purified
RUO
Yes
10044
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.