Learn More
serum/glucocorticoid regulated kinase 2, Mouse, Clone: 4G4, Abnova™
Mouse monoclonal antibody raised against a partial recombinant SGK2.
Brand: Abnova H00010110-M05.100ug
Description
This gene encodes a serine/threonine protein kinase. Although this gene product is similar to serum- and glucocorticoid-induced protein kinase (SGK), this gene is not induced by serum or glucocorticoids. This gene is induced in response to signals that activate phosphatidylinositol 3-kinase, which is also true for SGK. Two alternate transcripts encoding two different isoforms have been described. [provided by RefSeq
Sequence: SPINWDDLYHKRLTPPFNPNVTGPADLKHFDPEFTQEAVSKSIGCTPDTVASSSGASSAFLGFSYAPEDDDILDCSpecifications
| serum/glucocorticoid regulated kinase 2 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC065511 | |
| SGK2 | |
| SGK2 (AAH65511, 293 a.a. ∼ 367 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Yes | |
| 10110 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG3 κ |
| ELISA, Immunofluorescence, Western Blot | |
| 4G4 | |
| Mouse monoclonal antibody raised against a partial recombinant SGK2. | |
| SGK2 | |
| H-SGK2/dJ138B7.2 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Antibody |
Safety and Handling
- SGK2 monoclonal antibody (M05), clone 4G4
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P102-Keep out of reach of children.
- P103-Read label before use.
- P233-Keep container tightly closed.
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P404-Store in a closed container.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sodium phosphate dibasic, potassium phosphate monobasic
Your input is important to us. Please complete this form to provide feedback related to the content on this product.