Learn More
secreted frizzled-related protein 4, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SFRP4.
Brand: Abnova H00006424-A01.50uL
Description
Secreted frizzled-related protein 4 (SFRP4) is a member of the SFRP family that contains a cysteine-rich domain homologous to the putative Wnt-binding site of Frizzled proteins. SFRPs act as soluble modulators of Wnt signaling. The expression of SFRP4 in ventricular myocardium correlates with apoptosis related gene expression. [provided by RefSeq
Sequence: CNEVTTVVDVKEIFKSSSPIPRTQVPLITNSSCQCPHILPHQDVLIMCYEWRSRMMLLENCLVEKWRDQLSKRSIQWEERLQEQRRTVQDKKKTAGRTSRSNSpecifications
| secreted frizzled-related protein 4 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant SFRP4. | |
| SFRP4 | |
| FRP-4/FRPHE/MGC26498 | |
| Mouse | |
| 50 μL | |
| Membrane Receptors | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_003014.2 | |
| SFRP4 | |
| SFRP4 (NP_003005.1, 211 a.a. ∼ 312 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 6424 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.