missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse sFRP-3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£147.00 - £1420.00
Specifications
| Antigen | sFRP-3/FRZB |
|---|---|
| Clone | 133912 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18725138
![]() |
R&D Systems
MAB192-SP |
25 μg |
£147.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18171601
![]() |
R&D Systems
MAB192-050 |
50 μg |
£235.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18163431
![]() |
R&D Systems
MAB192-500 |
500 μg |
£1420.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
sFRP-3/FRZB Monoclonal specifically detects sFRP-3/FRZB in Human, Mouse samples. It is validated for Western Blot.Specifications
| sFRP-3/FRZB | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| AAB51298 | |
| 2487 | |
| Mouse myeloma cell line NS0-derived recombinant human sFRP-3 Ala33-Asn325 Accession # AAB51298 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human sFRP-3 in direct ELISAs and Western blots. Shows 100% cross-reactivity with recombinant mouse (rm) sFRP-3 and no cross-reactivity with recombinant human (rh) sFRP-1, rmsFRP-2, or rhsFRP-4. |
| 133912 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FIZ, FREOS1, Frezzled, Fritz, frizzled-related protein, Frizzled-related protein 1, FRP, FRP-3, FRZB, Frzb1, FrzB-1, FRZB1sFRP-3, FRZB-PEN, FZRB, hFIZ, secreted frizzled-related protein 3, sFRP3, SFRP3frezzled, SRFP3frizzled homolog-related | |
| FRZB | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title