missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SF3B14 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539354
This item is not returnable.
View return policy
Description
This gene encodes a 14 kDa protein subunit of the splicing factor 3b complex. Splicing factor 3b associates with both the U2 and U11/U12 small nuclear ribonucleoprotein complexes (U2 snRNP) of spliceosomes. This 14 kDa protein interacts directly with subunit 1 of the splicing factor 3b complex. This 14 kDa protein also interacts directly with the adenosine that carries out the first transesterification step of splicing at the pre-mRNA branch site. [provided by RefSeq, Jul 2008].
Specifications
| SF3B14 | |
| Polyclonal | |
| Unconjugated | |
| SF3B6 | |
| SAP14, SF3B14, SF3B14A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51639, 66055 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9Y3B4, P59708 | |
| SF3B6 | |
| A synthetic peptide derived from the C-terminal region of human SF3B14 | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction