missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SET domain containing (lysine methyltransferase) 7, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SET7.
Brand: Abnova H00080854-A01.50uL
This item is not returnable.
View return policy
Description
Sequence: SRDWALNGNTLSLDEETVIDVPEPYNHVSKYCASLGHKANHSFTPNCIYDMFVHPRFGPIKCIRTLRAVEADEELTVAYGYDHSPPGKSGPEAPEWYQVELKAFQATQQKSpecifications
| SET domain containing (lysine methyltransferase) 7 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant SET7. | |
| SETD7 | |
| FLJ21193/KIAA1717/KMT7/SET7/SET7/9/SET9 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 80854 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_030648 | |
| SETD7 | |
| SET7 (NP_085151, 257 a.a. ∼ 366 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human, Rat | |
| Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction