Learn More
anti-SERPINA6 Matched Antibody Pair, Abnova™
Brand: Abnova H00000866-AP11.1Set
Includes: Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-SERPINA6 (100μg; Detection antibody: mouse monoclonal anti-SERPINA6, IgG1 Kappa (20μg; Reagent are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
This gene encodes an α-globulin protein with corticosteroid-binding properties. This is the major transport protein for glucorticoids and progestins in the blood of most vertebrates. The gene localizes to a chromosomal region containing several closely related serine protease inhibitors which may have evolved by duplication events.
Host Species: Rabbit, Mouse
Isotype: IgG, IgGk1, κ
Cross Reactivity: Human
ELISA Pair
Specifications
| SERPINA6 | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human SERPINA6. | |
| CBG | |
| Mouse, Rabbit | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-SERPINA6 (100 μg) 2. Detection antibody: mouse monoclonal anti-SERPINA6, IgG1 Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| ELISA | |
| Unconjugated | |
| SERPINA6 | |
| SERPINA6 | |
| 1 Set | |
| 866 | |
| Human | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.