Learn More
septin 11, Mouse, Clone: 1E5, Abnova™
Mouse monoclonal antibody raised against a partial recombinant SEPT11.
Brand: Abnova H00055752-M02.100ug
Description
SEPT11 belongs to the conserved septin family of filament-forming cytoskeletal GTPases that are involved in a variety of cellular functions including cytokinesis and vesicle trafficking (Hanai et al., 2004 [PubMed 15196925]; Nagata et al., 2004 [PubMed 15485874]).[supplied by OMIM
Sequence: LRNLSLSGHVGFDSLPDQLVNKSTSQGFCFNILCVGETGIGKSTLMDTLFNTKFESDPATHNEPGVRLKARSYELQESNVRLKLTIVDTVGFGDQINKDDSYKPIVEYIDSpecifications
| Septin-11 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC008083 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 55752 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b λ |
| ELISA, Western Blot | |
| 1E5 | |
| Mouse monoclonal antibody raised against a partial recombinant SEPT11. | |
| SEPT11 | |
| SEPT11 | |
| SEPT11 (AAH08083, 71 a.a. ∼ 180 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Cell Cycle | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.