Learn More
SENP8, Human, IP-WB Antibody Pair (PW3), Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00123228-PW3.1Set
Product Code. 16113844
  • £729.00 / 1 set
Includes: Antibody pair for IP: rabbit polyclonal anti-SENP8 (300μL), Antibody pair for WB: mouse monoclonal anti-SENP8 (50μg)
Specifications
SENP8
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
DEN1/HsT17512/NEDP1/PRSC2
Mouse, Rabbit
RUO
123228
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
SENP8
SENP8
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-SENP8 (300 ul) 2. Antibody pair for WB: mouse monoclonal anti-SENP8 (50 ug)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.