missing translation for 'onlineSavingsMsg'
Learn More
Learn More
sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4D, Mouse, Clone: 3B4, Abnova™
Mouse monoclonal antibody raised against a partial recombinant SEMA4D.
Brand: Abnova H00010507-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: LQPLSATSLYVCGTNAFQPACDHLNLTSFKFLGKNEDGKGRCPFDPAHSYTSVMVDGELYSGTSYNFLGSEPIISRNSSHSPLRTEYAIPWLNEPSFVFADVIRKSPDSPSpecifications
| sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4D | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC054500 | |
| SEMA4D | |
| SEMA4D (AAH54500, 115 a.a. ∼ 224 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Apoptosis | |
| 10507 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
| ELISA, Western Blot | |
| 3B4 | |
| Mouse monoclonal antibody raised against a partial recombinant SEMA4D. | |
| SEMA4D | |
| C9orf164/CD100/FLJ33485/FLJ34282/FLJ39737/FLJ46484/M-sema-G/MGC169138/MGC169141/SEMAJ/coll-4 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction