Learn More
SEMA4A, Mouse, Clone: 4E2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant SEMA4A.
Brand: Abnova H00064218-M02.100ug
Additional Details : Weight : 0.00010kg
Description
SEMA4A is a member of the semaphorin family of soluble and transmembrane proteins. Semaphorins are involved in guidance of axonal migration during neuronal development and in immune responses.[supplied by OMIM
Sequence: GGGQGPMPRVRYYAGDERRALSFFHQKGLQDFDTLLLSGDGNTLYVGAREAILALDIQDPGVPRLKNMIPWPASDRKKSECAFKKKSNETQCFNFIRVLVSpecifications
SEMA4A | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_022367 | |
SEMA4A | |
SEMA4A (NP_071762, 33 a.a. ∼ 132 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA | |
4E2 | |
Mouse monoclonal antibody raised against a partial recombinant SEMA4A. | |
SEMA4A | |
CORD10/FLJ12287/RP35/SEMAB/SEMB | |
Mouse | |
Affinity chromatography | |
RUO | |
64218 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |