missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SEMA3C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA524997
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine based on sequence homology.
The SEMA3C gene encodes the protein semaphorin 3C, a member of the semaphorin family known for its roles in axonal guidance, cellular signaling, and vascular development. Semaphorin 3C is involved in guiding neurons during development by serving as a repulsive cue, helping establish proper neural network formation through interaction with neuropilin and plexin receptors. It also plays crucial roles in angiogenesis and cardiac development by influencing endothelial cell behavior and migration. In the context of cancer, SEMA3C is implicated in tumor progression, where its dysregulation can promote tumor cell invasion and metastasis by modifying the extracellular matrix and affecting cellular adhesion. Research has demonstrated that SEMA3C can alter the tumor microenvironment, making it a potential target for cancer therapies aimed at inhibiting metastasis and controlling tumor growth. Understanding SEMA3C's function in various biological processes continues, highlighting its significance in developmental biology and disease modulation.
Specifications
| SEMA3C | |
| Polyclonal | |
| Unconjugated | |
| SEMA3C | |
| 1110036B02Rik; sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3C; sema E; Sema3c; Semae; semaphorin 3C; semaphorin E; semaphorin-3C; Semaphorin-E; SemE | |
| Rabbit | |
| Protein A, Antigen affinity chromatography | |
| RUO | |
| 10512, 20348 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.37 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q62181, Q99985 | |
| SEMA3C | |
| KLH conjugated synthetic peptide between 592-620 amino acids from the C-terminal region of human SEMA3C. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction