missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SECTM1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA542725
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: ARDSHAGLYM WHLVGHQRNN RQVTLEVSGA EPQSAPDTGF WPVPAVVTAV Sequence homology: Human: 100%
SECTM1 Polyclonal antibody specifically detects SECTM1 in Human samples. It is validated for Western Blot.Specifications
SECTM1 | |
Polyclonal | |
Unconjugated | |
SECTM1 | |
1810003C24Rik; K12; Protein K-12; secreted and transmembrane 1; secreted and transmembrane 1B; secreted and transmembrane protein 1; secreted and transmembrane protein 1b; SECTM1; Sectm1b; type 1a transmembrane protein | |
Rabbit | |
Affinity Chromatography | |
RUO | |
6398 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8WVN6 | |
SECTM1 | |
Synthetic peptide directed towards the middle region of human SECTM1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |