missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCTR Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539706
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a G protein-coupled receptor and belongs to the glucagon-VIP-secretin receptor family. It binds secretin which is the most potent regulator of pancreatic bicarbonate, electrolyte and volume secretion. Secretin and its receptor are suggested to be involved in pancreatic cancer and autism. [provided by RefSeq, Jul 2008].
Specifications
| SCTR | |
| Polyclonal | |
| Unconjugated | |
| SCTR | |
| SCTR | |
| A synthetic peptide derived from the internal region of human SCTR | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P47872 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6344 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction