missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCN3B Polyclonal Antibody

SCN3B Polyclonal Antibody
Brand: Invitrogen PA541403
This item is not returnable.
View return policy
Description
SCN3B Polyclonal Antibody for Western Blot
Specifications
| SCN3B | |
| Polyclonal | |
| Unconjugated | |
| Scn3b | |
| 1110001K16Rik; 4833414B02Rik; ATFB16; brain and heart sodium channel beta 3 subunit; BRGDA7; HSA243396; KIAA1158; LOC540925 protein; Scn3b; Scnb3; sodium channel beta 3 subunit; sodium channel subunit beta-3; sodium channel, voltage gated, type III beta subunit; sodium channel, voltage-gated, type III, beta; sodium channel, voltage-gated, type III, beta subunit; sodium voltage-gated channel beta subunit 3; voltage-gated sodium channel beta-3 subunit | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 55800 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NY72 | |
| Scn3b | |
| Synthetic peptide directed towards the N-terminal of human SCN3B (aa 65-114). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur