Learn More
Mammaglobin A Monoclonal Antibody (3C8), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515716
Code : A0
Additional Details : Weight : 0.01000kg
Description
MA5-15716 targets SCGB2A2 in indirect ELISA, FACS, IF, IHC, and WB applications and shows reactivity with Human samples. The MA5-15716 immunogen is purified recombinant fragment of human SCGB2A2 expressed in E. Coli.<br>. MA5-15716 detects SCGB2A2 which has a predicted molecular weight of approximately 11kDa.
Mammaglobin A Monoclonal antibody specifically detects Mammaglobin A in Human samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot.Specifications
Mammaglobin A | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
Q13296 | |
SCGB2A2 | |
Purified recombinant fragment of human SCGB2A2 expressed in E. Coli. | |
RUO | |
4250 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot | |
3C8 | |
Unconjugated | |
SCGB2A2 | |
C3.1; HGNC:7050; mammaglobin 1; mammaglobin A; Mammaglobin-1; Mammaglobin-A; MGB1; MGC71974; prostatein; prostatein C3 subunit; prostatein peptide C3; prostatic steroid binding protein, allele C3(1); prostatic steroid-binding protein C3; Psbpc3; SCGB2A2; Secretoglobin family 2A member 2; secretoglobin, family 2A, member 2; UGB2 | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |