Learn More
secretogranin II (chromogranin C), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SCG2.
Brand: Abnova H00007857-A01.50uL
Description
The protein encoded by this gene is a member of the chromogranin/secretogranin family of neuroendocrine secretory proteins. Studies in rodents suggest that the full-length protein, secretogranin II, is involved in the packaging or sorting of peptide hormones and neuropeptides into secretory vesicles. The full-length protein is cleaved to produce the active peptide secretoneurin, which exerts chemotaxic effects on specific cell types, and EM66, whose function is unknown. [provided by RefSeq
Sequence: LARMLVKYPEIINSNQVKRVPGQGSSEDDLQEEEQIEQAIKEHLNQGSSQETDKLAPVSKRFPVGPPKNDDTPNRQYWDEDLLMKVLEYLNQEKAEKGREHIAKRAMENMSpecifications
| secretogranin II (chromogranin C) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant SCG2. | |
| SCG2 | |
| CHGC/SN/SgII | |
| Mouse | |
| 50 μL | |
| Angiogenesis | |
| 7857 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_003469 | |
| SCG2 | |
| SCG2 (NP_003460, 508 a.a. ∼ 617 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.