missing translation for 'onlineSavingsMsg'
Learn More

Proteintech™ Anti-SARS-CoV-2 S-RBD Protein Human IgG ELISA Kit

Product Code. 16851656 Shop All Proteintech Products
Change view
Click to view available options
Immunoassay Kit Format:
Coated ELISA Kit
Unit Size:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Immunoassay Kit Format Unit Size
16851656 Coated ELISA Kit 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16851656 Supplier Proteintech™ Supplier No. KE3000396T
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Quantitatively measures human IgG for 2019-nCoV s-RBD in serum and plasma

  • Indirect ELISA kit
  • S-RBD Recombinant Protein is pre-coated onto microplate well
  • Samples or standard added to well
  • After incubation, wells are washed and horseradish peroxidase conjugated anti-Human IgG is added to each well
    • Produces complex Recombinant Protein-human anti-S-RBD IgG antibody-HRP conjugated antibody
  • After incubation, wells are washed followed by Tetramethyl-benzidine (TMB) reagent
  • Solution containing sulfuric acid used to stop color development and color intensity
    • Proportional to quantity of bound protein, measurable at 450 nm with correction wavelength set at 630 nm

Specifications

Accession Number NC_045512
Assay Range 6.25 to 200 ng/mL
Certifications/Compliance ISO 13485
Description Tests: 1 x 96 well plate
Assay Indirect
Immunoassay Kit Format Coated ELISA Kit
Sample Type Plasma, Serum
Substrate DAB
Immunoassay Kit Grade Mix and Match Panel
Interassay CV <10%
Intraassay CV <10%
Shipping Condition Blue ice
Storage Requirements -20°C to +4°C
Total Assay Time 1.5 hr.
UniProt ID P0DTC2
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 96TEST Anti-SARS-CoV-2 S-RBD protein Human IgG E
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Sulphuric Acid, Tetramethylbenzidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.