missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Anti-SARS-CoV-2 Neutralizing Antibody Titer Serologic Assay Kit

Product Code. 17246570
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests

Product Code. 17246570

Brand: ACROBiosystems TASK02296tests

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

The newly identified 2019 novel coronavirus (SARS-CoV-2) has posed a serious threat to human health, and it spreads quickly. It is helpful to develop the Anti SARS-CoV-2 neutralizing antibody titer serologic assay kit to test the protective neutralizing antibody level in serum.

Specifications

Product Type Antibody titer assay kits
Quantity 96 Tests
Shipping Condition RT
Storage Requirements 2°C to 8°C
Target RBD
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.