missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Anti-SARS-CoV-2 Antibody IgG Titer Serologic Assay kit (Nucleocapsid Protein)

Product Code. 17196791
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests

Product Code. 17196791

Brand: ACROBiosystems TASK00896tests

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

kit employs a standard indirect-ELISA format, providing a rapid detection of Anti-SARS-CoV-2 antibodies in serum

Nucleocapsid (N) protein is the most abundant protein found in coronavirus. CoV N protein is a highly immunogenic phosphoprotein important for viral genome replication and modulation of cell signaling pathways. It was first identified by a research team while they were screening for ADP-ribosylated proteins during coronavirus (CoV) infection (Grunewald M. E., et al. 2017, Virology; 517: 62-68). The array of diverse functional activities accommodated in N protein makes it more than a structural protein but also an interesting target in the development of antiviral therapeutics. Because of the conservation of N protein sequence and its strong immunogenicity, N protein of coronavirus is chosen as a diagnostic tool.

Specifications

Product Type Antibody titer assay kits
Kit Contents This assay kit employs a standard indirect-ELISA format, providing a rapid detection of Anti-SARS-CoV-2 Nucleocapsid Antibody, human IgG in serum by SARS-CoV-2 Nucleocapsid Protein. The kit consists of High-bind Plate, Nucleocapsid Protein, an Anti-SARS-C
Quantity 96 Tests
Shipping Condition RT
Storage Requirements -20°C, aviod light
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.