missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha Sarcoglycan Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA537050
Code : NEW
Additional Details : Weight : 0.00910kg
Description
This antibody detects endogenous protein at a molecular weight of 50 kDa. Purity is >95% by SDS-PAGE.
Alpha Sarcoglycan Polyclonal antibody specifically detects Alpha Sarcoglycan in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blot.Specifications
Alpha Sarcoglycan | |
Polyclonal | |
Unconjugated | |
SGCA | |
50 kDa dystrophin-associated glycoprotein; 50DAG; 50kD DAG; adhalin; ADL; Alpha-sarcoglycan; alpha-SG; Asg; DAG2; DMDA2; Dystroglycan 2; Dystroglycan2; dystroglycan-2; LGMD2D; limb girdle muscular dystrophy 2D; sarcoglycan alpha; sarcoglycan, alpha (50kDa dystrophin-associated glycoprotein); sarcoglycan, alpha (dystrophin-associated glycoprotein); SCARMD1; SG alpha; SG-alpha; Sgca | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20391, 303468, 6442 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P82350, Q16586 | |
SGCA | |
Synthetic peptide corresponding to amino acids 209-254 of human Sarcoglycan-alpha | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |