Learn More
S100P Mouse anti-Human, Unlabeled, Clone: 16, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 610307
Description
The S100 multigene family consists of a number of small dimeric molecules around 100 residues in length. The original members of this family, S100α and S100β, were initially identified in bovine brain. S100P is a novel member of this family which was isolated from human placenta. The S100 family consists of at least 10 members which show a cell-type-specific expression pattern. Most S100 proteins are believed to be involved in cytoplasmic Ca2+ control. S100P consists of 95 amino acids and shares about 50% sequence identity with S100α and S100α. Like other S100 proteins, S100P functions as a homodimer and binds two Ca2+ ions per monomer. Each S100P molecule contains both a low affinity and a high affinity Ca2+ binding site.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
| S100P | |
| Monoclonal | |
| 250μg/mL | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| 150 μg | |
| Cell Biology | |
| Human | |
| IgG1 |
| Western Blot | |
| 16 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.