Learn More
retinoid X receptor, alpha, Mouse, Clone: 4B5, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant RXRA.
Brand: Abnova H00006256-M03.100ug
Description
Retinoid X receptors (RXRs) and retinoic acid receptors (RARs), are nuclear receptors that mediate the biological effects of retinoids by their involvement in retinoic acid-mediated gene activation. These receptors exert their action by binding, as homodimers or heterodimers, to specific sequences in the promoters of target genes and regulating their transcription. The protein encoded by this gene is a member of the steroid and thyroid hormone receptor superfamily of transcriptional regulators. [provided by RefSeq
Sequence: MLGSWPARTFHPGACVSRRPSAPWKHTASGKDSPDLRFSEHGVSQEFWAGGLVAVLEMTPSPSPWGTQEGPAGMCSLWVVGWCPCRGAGVRDLVLVHAGVWCKHVCAVQRDACGESRTPAPPRKGGAVTSVLCLFLIKTFPLFSYKFASCKQVHKDPPLVKSGFESpecifications
| retinoid X receptor, alpha | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC007925 | |
| RXRA | |
| RXRA (AAH07925, 1 a.a. ∼ 165 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Transcription Regulation | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunoprecipitation, Western Blot | |
| 4B5 | |
| Mouse monoclonal antibody raised against a full length recombinant RXRA. | |
| RXRA | |
| FLJ00280/FLJ00318/FLJ16020/FLJ16733/MGC102720/NR2B1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 6256 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.