Learn More
RUNX2, Mouse, Clone: 4C8, Abnova™
Mouse monoclonal antibody raised against a partial recombinant RUNX2.
Brand: Abnova H00000860-M07.100ug
Description
This gene is a member of the RUNX family of transcription factors and encodes a nuclear protein with an Runt DNA-binding domain. This protein is essential for osteoblastic differentiation and skeletal morphogenesis and acts as a scaffold for nucleic acids and regulatory factors involved in skeletal gene expression. The protein can bind DNA both as a monomer or, with more affinity, as a subunit of a heterodimeric complex. Mutations in this gene have been associated with the bone development disorder cleidocranial dysplasia (CCD). Transcript variants that encode different protein isoforms result from the use of alternate promoters as well as alternate splicing. [provided by RefSeq
Sequence: NPRPSLNSAPSPFNPQGQSQITDPRQAQSSPPWSYDQSYPSYLSQMTSPSIHSTTPLSSTRGTGLPAITDVPRRISDDDTATSDFCLWPSTLSKKSQAGASpecifications
| RUNX2 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_004348 | |
| RUNX2 | |
| RUNX2 (NP_004339, 251 a.a. ∼ 350 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG2a κ |
| ELISA, Immunofluorescence, Western Blot | |
| 4C8 | |
| Mouse monoclonal antibody raised against a partial recombinant RUNX2. | |
| RUNX2 | |
| AML3/CBFA1/CCD/CCD1/MGC120022/MGC120023/OSF2/PEA2aA/PEBP2A1/PEBP2A2/PEBP2aA/PEBP2aA1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 860 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.