missing translation for 'onlineSavingsMsg'
Learn More
Learn More
reticulon 3, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human RTN3 protein.
Brand: Abnova H00010313-B01P.50ug
This item is not returnable.
View return policy
Description
The reticulons are a group of highly conserved genes with preferential expression in neuroendocrine tissues (see, e.g., RTN1; MIM 600865).[supplied by OMIM
Sequence: MAEPSAATQSHSISSSSFGAEPSAPGGGGSPGACPALGTKSCSSSCAVHDLIFWRDVKKTGFVFGTTLIMLLSLAAFSVISVVSYLILALLSVTISFRIYKSVIQAVQKSEEGHPFKAYLDVDITLSSEAFHNYMNAAMVHINRALKLIIRLFLVEDLVDSLKLAVFMWLMTYVGAVFNGITLLILAELLIFSVPIVYEKYKTQIDHYVGIARDQTKSIVEKIQAKLPGIAKKKAESpecifications
| reticulon 3 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human RTN3 protein. | |
| RTN3 | |
| ASYIP/HAP/NSPL2/NSPLII/RTN3-A1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_006054.2 | |
| RTN3 | |
| RTN3 (NP_006045.1, 1 a.a. ∼ 236 a.a) full-length human protein. | |
| 50 μg | |
| Apoptosis | |
| 10313 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction