missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rtf1, Paf1/RNA polymerase II complex component, homolog (S. cerevisiae), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human RTF1 protein.
Brand: Abnova H00023168-B01.50uL
This item is not returnable.
View return policy
Description
Sequence: MKKQANKTASSGSSDKDSSAESSAPEEGEVSDSDSNSSSSSSDSDSSSEDEEFHDGYGEDLMGDEEDRARLEQMTEKEREQELFNRIEKREVLKRRFEIKKKLKTAKKKEKKEKKKKQEEEQEKKKLTQIQESQVTSHNKERRSKRDEKLDKKSQAMEELKAEREKRKNRTAELLAKKQPLKTSEVYSDDEEEEEDDKSSEKSDRSSRTSSSDEEEEKEEIPPKSQPVSLPEELNRVRLSRHKLERWCHMPFFAKTVTGCFVRIGIGNHNSKPVYRVAEITGVVETAKVYQLGGTRTNKGLQLRHGNDQRVFRLEFVSNQEFTESEFMKWKEAMFSAGMQLPTLDEINKKELSIKEALNYKFNDQDIEEIVKEKERFRKAPPNYAMKKTQLLKEKAMAEDLGDQDKAKQIQDQLNELEERAEALDRQRTKNISAISYINQRNREWNIVESEKALVAESHNMKNQQMDPFTRRQCKPTIVSNSRDPAVQAAILAQLNAKYGSGVLPDAPKEMSKGQGKDKDLNSKSASDLSEDLFKVHDFDVKIDLQVPSSESKALAITSKAPPAKDGAPRRSLNLEDYKKRRGLISpecifications
| Rtf1, Paf1/RNA polymerase II complex component, homolog (S. cerevisiae) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human RTF1 protein. | |
| RTF1 | |
| CDG1N/GTL7/KIAA0252 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 23168 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| No additive | |
| NM_015138.3 | |
| RTF1 | |
| RTF1 (NP_055953.1, 1 a.a. ∼ 585 a.a) full-length human protein. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction