Learn More
RT1B Mouse anti-Rat, PerCP, Clone: OX-6, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 557016
Description
The OX-6 antibody reacts with non-polymorphic determinants of the Rat MHC class II antigen, I-A equivalent. RT1B is found on peripheral B lymphocytes, thymic cortical epithelial and medullary reticular cells, small intestinal villus epithelium, epidermal Langerhans cells, dendritic cells, some tissue macrophage populations, peritoneal mast cells, and a subset of thymocytes, but not on peripheral T cells, erythrocytes, or microglia. The OX-6 mAb cross-reacts with mouse I-A[k] and I-A[s] alloantigens and with a major subset of splenocytes from NOD (I-A[g7]) mice. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: OX-6
Isotype: IgG1, κ
Cross Reactivity: Rat
Flow Cytometry
Specifications
| RT1B | |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Ia-like Glycoproteins from Wistar Thymocytes | |
| 0.1 mg | |
| Immunology | |
| Mouse, Rat | |
| IgG1 κ |
| Flow Cytometry | |
| OX-6 | |
| PerCP | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.