Learn More
ribosomal protein S9, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant RPS9.
Brand: Abnova H00006203-A01.50uL
Description
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S4P family of ribosomal proteins. It is located in the cytoplasm. Variable expression of this gene in colorectal cancers compared to adjacent normal tissues has been observed, although no correlation between the level of expression and the severity of the disease has been found. As is typical for genes encoding ribosomal proteins, multiple processed pseudogenes derived from this gene are dispersed through the genome. [provided by RefSeq
Sequence: FLERRLQTQVFKLGLAKSIHHARVLIRQRHIRVRKQVVNIPSFIVRLDSQKHIDFSLRSPYGGGRPGRVKRKNAKKGQGGAGAGDDEEEDSpecifications
| ribosomal protein S9 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant RPS9. | |
| RPS9 | |
| RPS9 | |
| RPS9 (NP_001004, 105 a.a. ∼ 194 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human, Rat | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_001013 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 6203 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.