missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RPA70 Monoclonal Antibody (4C4)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA517166

Product Code. 15318120

  • £437.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MA5-17166 targets RPA1 in indirect ELISA, FACS, ICC, IHC, IF and WB applications and shows reactivity with Human and Non-human primate samples. The MA5-17166 immunogen is purified recombinant fragment of human RPA1 expressed in E. Coli. MA5-17166 detects RPA1 which has a predicted molecular weight of approximately 68.1kDa.

This gene Plays an essential role in several cellular processes in DNA metabolism including replication, recombination and DNA repair. Binds and subsequently stabilizes single-stranded DNA intermediates and thus prevents complementary DNA from reannealing.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RPA70
Monoclonal
1 mg/mL
PBS with 0.05% sodium azide
P27694
RPA1
Purified recombinant fragment of human RPA1 expressed in E. Coli.
100 μg
Primary
Human, Monkey
Antibody
IgG1
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
4C4
Unconjugated
RPA1
5031405K23Rik; 70kDa; AA589576; AW557552; Cb1-727; HSSB; MST075; MSTP075; REPA1; Replication factor A protein 1; replication protein A 70 kDa DNA-binding subunit; Replication protein A 70 kDa DNA-binding subunit, N-terminally processed; replication protein A1; replication protein A1, 70kDa; RF-A; RF-A protein 1; Rpa; RP-A; RP-A p70; RPA1; RPA70; Single-stranded DNA-binding protein
Mouse
Protein G
RUO
6117
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.