missing translation for 'onlineSavingsMsg'
Learn More

RORγt Mouse anti-Mouse, PE, Clone: Q31-378, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 562607

Product Code. 15892069

  • ONLINE EXCLUSIVE PRICE
    £251.00
    £247.59 / Each
    Save £3.41
    1% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The Q31-378 monoclonal antibody recognizes RORgamma t (RORγt), an isoform of RORgamma (RORγ). RORγt is a DNA-binding transcription factor that belongs to the ROR/RZR orphan nuclear receptor family. RORγt is expressed exclusively by lymphoid cells including CD4+CD8+ thymocytes, peripheral CD4+ Th17 and CD8+ Tc17 cells, NKT cells and innate lymphoid cells such as lymphoid tissue inducer (LTi) cells. RORγt plays essential roles in thymopoiesis, T cell homeostasis, differentiation of effector T lymphocytes and the development of secondary lymphoid tissues including lymph nodes and Peyer's patches.

Host Species: Mouse
Clone: Q31-378
Isotype: IgG2a, κ
Species Reactivity: Mouse
Immunogen: Mouse RORγt Recombinant Protein

Intracellular Staining

TRUSTED_SUSTAINABILITY
Specifications

Specifications

RORγt
Monoclonal
0.2mg/mL
RORγT; RORgt; RORgamma t; RORgammaT; Rorc2; Rorg; TOR; Thor; Nr1f3
Mouse
Affinity Purified
RUO
Mouse
IgG2a κ
Flow Cytometry
Q31-378
PE
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse RORγt Recombinant Protein
50 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.