missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse ROR gamma t/RORC2/NR1F3 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
£237.00 - £454.00
Specifications
| Antigen | ROR gamma t/RORC2/NR1F3 |
|---|---|
| Clone | 600380 |
| Dilution | Intracellular Staining by Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18742667
![]() |
R&D Systems
IC6006P-025 |
25 Tests |
£237.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18714237
![]() |
R&D Systems
IC6006P-100 |
100 Tests |
£454.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ROR gamma t/RORC2/NR1F3 Monoclonal specifically detects ROR gamma t/RORC2/NR1F3 in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| ROR gamma t/RORC2/NR1F3 | |
| Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| 6097 | |
| Human ROR gamma t peptide Met1-Arg10 Accession # P51449 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| 600380 | |
| Flow Cytometry | |
| R-PE | |
| Mouse | |
| Human, Mouse | |
| P51449 | |
| RORC | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human and mouse ROR gamma t/RORC2 in direct ELISAs. In direct ELISAs, no cross-reactivity with human and mouse RORC. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title