missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROR gamma Polyclonal Antibody, Invitrogen™
ROR gamma Antibody, PA5-28257, from Invitrogen™. Species Reactivity: Human; Applications: Western Blot.
Brand: Invitrogen PA528257
This item is not returnable.
View return policy
Description
ROR gamma Antibody, PA5-28257, from Invitrogen. Species Reactivity: Human; Applications: Western Blot.
The protein encoded by this gene is a DNA-binding transcription factor and is a member of the NR1 subfamily of nuclear hormone receptors. The specific functions of this protein are not known; however, studies of a similar gene in mice have shown that this gene may be essential for lymphoid organogenesis and may play an important regulatory role in thymopoiesis. In addition, studies in mice suggest that the protein encoded by this gene may inhibit the expression of Fas ligand and IL2. Two transcript variants encoding different isoforms have been found for this gene.Host Species: Rabbit
Cross Reactivity: Human
Isotype: IgG
Immunogen: Recombinant fragment corresponding to a region within amino acids 95 and 518 of Human ROR gamma
Formulation: 100μL purified antibody (0.72mg/mL) in 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Synonyms: NR1F3, RORG, RZR-GAMMA, RZRG, TOR, RAR-related orphan nuclear receptor variant 2, nuclear receptor ROR-gamma, nuclear receptor RZR-gamma, nuclear receptor subfamily 1 group F member 3, retinoic acid-binding receptor gamma, retinoid-related orphan receptor gamma
Uniprot Accession: P51449
Entrez GeneID: 6097
Gene Symbol: RORC
PA5-28257 targets ROR gamma in WB applications and shows reactivity with Human samples. The PA5-28257 immunogen is recombinant fragment corresponding to a region within amino acids 95 and 518 of Human ROR gamma.
Specifications
| ROR gamma | |
| Polyclonal | |
| P51449 | |
| RORC | |
| Recombinant fragment corresponding to a region within amino acids 95 and 518 of Human ROR gamma | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| NR1F3, RORG, RZR-GAMMA, RZRG, TOR, RAR-related orphan nuclear receptor variant 2, nuclear receptor ROR-gamma, nuclear receptor RZR-gamma, nuclear receptor subfamily 1 group F member 3, retinoic acid-binding receptor gamma, retinoid-related orphan receptor gamma | |
| Rabbit | |
| 100 μL | |
| 6097 | |
| Store at -20°C, Avoid Freeze/Thaw Cycles | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction