Learn More
ROCK-II Mouse, Unlabeled, Clone: 21, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 610623
Description
Activity of the GTP-binding proteins is regulated by GAPs, that accelerate binding, and GTPases, that enhance the rate of GTP hydrolysis (Ras, Rho, cdc42Hs, and Rac). The isoforms of Rho, a small GTP-binding protein, regulate cellular processes such as the formation of the stress fibers, lamelipodia, and filopodia. ROKα (RhoA-binding kinase) is a Ser/Thr protein kinase that interects with the GTP-binding form of RhoA. Like RhoA, ROKα localizes at the cellular margins and colocalizes with acti filaments. The ROKα gene encodes a protein of 1302 amino acids with homology to the human myotonic kinase. Although ROKα binds to GTP-RhoA, it is not yet clear whether this interaction induces the kinase activity of ROKα.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
| ROCK-II | |
| Monoclonal | |
| 250μg/mL | |
| ROKα | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
| Western Blot | |
| 21 | |
| Unconjugated | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Rat ROKα aa. 567-718 | |
| 50 μg | |
| Cell Biology | |
| Canine, Human, Mouse, Rat | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.