missing translation for 'onlineSavingsMsg'
Learn More
Learn More
roundabout, axon guidance receptor, homolog 3 (Drosophila), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human ROBO3 protein.
Brand: Abnova H00064221-B02.50uL
This item is not returnable.
View return policy
Description
Sequence: MTPPLQGPRARFRKKPKALPYRRENSPGDLPPPPLPPPEEEASWALELRAAGSMSSLERERSGERKAVQAVPLAAQRVLHPDEEAWLPYSRPSFLSRGQGTSTCSTAGSNSSRGSSSSRGSRGPGRSRSQSRSQSQRPGQKRREEPRSpecifications
| roundabout, axon guidance receptor, homolog 3 (Drosophila) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human ROBO3 protein. | |
| ROBO3 | |
| FLJ21044/HGPPS/HGPS/RBIG1/RIG1 | |
| Mouse | |
| 50 μL | |
| Neurobiology | |
| Primary | |
| Human | |
| Serum |
| Western Blot | |
| Unconjugated | |
| No additive | |
| BC008623 | |
| ROBO3 | |
| ROBO3 (AAH08623, 1 a.a. ∼ 147 a.a) full-length human protein. | |
| RUO | |
| Yes | |
| 64221 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction