Learn More
ring finger protein 8, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human RNF8 protein.
Brand: Abnova H00009025-B01.50uL
Description
The protein encoded by this gene contains a RING finger motif and a FHA domain. This protein has been shown to interact with several class II ubiquitin-conjugating enzymes (E2), including UBE2E1/UBCH6, UBE2E2, and UBE2E3, and may act as an ubiquitin ligase (E3) in the ubiquitination of certain nuclear proteins. Alternatively spliced transcript variants encoding distinct isoforms have been reported. [provided by RefSeq
Sequence: MGEPGFFVTGDRAGGRSWCLRRVGMSAGWLLLEDGCEVTVGRGFGVTYQLVSKICPLMISRNHCVLKQNPEGQWTIMDNKSLNGVWLNRARLEPLRVYSIHQGDYIQLGVPLENKENAEYEYEVTEEDWETIYPCLSPKNDQMIEKNKELRTKRKFSLDELAGPGAEGPSNLKSKINKVSCESGQPVKSQGKGEVASTPSDNLDPKLTALEPSKTTGAPIYPGFPKVTEVHHEQKASNSSASQRSLQMFKVTMSRILRLKIQMQEKHEAVMNVKKQTQKGNSKKVVQMEQELQDLQSQLCAEQAQQQARVEQLEKTFQEEEQHLQGLEIAQGEKDLKQQLAQALQEHWALMEELNRSKKDFEAIIQAKNKELEQTKEEKEKMQAQKEEVLSHMNDVLENELQCIICSEYFIEAVTLNCAHSFCSYCINEWMKRKIECPICRKDIKSKTYSLVLDNCINKMVNNLSSEVKERRIVLIRERKAKRLFSpecifications
| ring finger protein 8 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human RNF8 protein. | |
| RNF8 | |
| FLJ12013/KIAA0646 | |
| Mouse | |
| 50 μL | |
| Stem Cell Biology | |
| Primary | |
| Human | |
| Serum |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| No additive | |
| NM_003958.2 | |
| RNF8 | |
| RNF8 (NP_003949.1, 1 a.a. ∼ 485 a.a) full-length human protein. | |
| RUO | |
| Yes | |
| 9025 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.