Learn More
ring finger protein 19A, Mouse, Clone: 4E4, Abnova™
Mouse monoclonal antibody raised against a partial recombinant RNF19.
Brand: Abnova H00025897-M01A.200uL
Description
The protein encoded this gene contains two RING-finger motifs and an IBR (in between RING fingers) motif. This protein is an E3 ubiquintin ligase that is localized in Lewy bodies (LBs), a characteristic neuronal inclusion in Parkinson's disease (PD) brains. This protein interacts with UBE2L3/UBCH7 and UBE2E2/UBCH8, but not other ubiquitin-conjugating enzymes. This protein is found to bind and ubiquitylate synphilin 1 (SNCAIP), which is a interacting protein of alpha synuclein in neurons, and a major component of LB. Alternatively spliced transcript variants encoding the same protein have been reported. [provided by RefSeq]
Sequence: MKTSCSHGSSDYHTRFATVNILPEVENDRLENSPHQCSISVVTQTASCSEVSQLNHIAEEHGNNGIKPNVDLYFGDALKETNNNHSHQTMELKVAIQTESpecifications
| ring finger protein 19A | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_015435 | |
| RNF19A | |
| RNF19 (NP_056250, 739 a.a. ∼ 837 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| Yes | |
| 25897 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 4E4 | |
| Mouse monoclonal antibody raised against a partial recombinant RNF19. | |
| RNF19A | |
| DKFZp566B1346/DORFIN/RNF19 | |
| Mouse | |
| 200 μL | |
| Ubiquitin/Proteasome | |
| Primary | |
| Human | |
| Antibody | |
| IgM κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.