missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ring finger protein 182, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant RNF182.
Brand: Abnova H00221687-A01.50uL
This item is not returnable.
View return policy
Description
Sequence: MASQPPEDTAESQASDELECKICYNRYNLKQRKPKVLECCHRVCAKCLYKIIDFGDSPQGVIVCPFCRFETCLPDDEVSSLPDDNNILVNLTCGGKGKKSpecifications
| ring finger protein 182 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant RNF182. | |
| RNF182 | |
| FLJ40772/MGC33993 | |
| Mouse | |
| 50 μL | |
| Ubiquitin/Proteasome | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_152737 | |
| RNF182 | |
| RNF182 (NP_689950, 1 a.a. ∼ 99 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 221687 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction