missing translation for 'onlineSavingsMsg'
Learn More

ring finger protein 135, Mouse, Polyclonal Antibody, Abnova™

Product Code. 16160707
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16160707 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16160707 Supplier Abnova Supplier No. H00084282B01.50uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse polyclonal antibody raised against a full-length human RNF135 protein.

The protein encoded by this gene contains a RING finger domain, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. This gene is located in a chromosomal region known to be frequently deleted in patients with neurofibromatosis. Alternatively spliced transcript variants encoding distinct isoforms have been reported. [provided by RefSeq

Sequence: MAGLGLGSAVPVWLAEDDLGCIICQGLLDWPATLPCGHSFCRHCLEALWGARDARRWACPTCRQGAAQQPHLRKNTLLQDLADKYRRAAREIQAGSDPAHCPCPGSSSLSSAAARPRRRPELQRVAVEKSITEVAQELTELVEHLVDIVRSLQNQRPLSESGPDNELSILGKENSWKPRLPPHAHCLTRATLHSGELLGLLSGPSIQPLT

Specifications

Antigen ring finger protein 135
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human RNF135 protein.
Formulation No additive
Gene RNF135
Gene Accession No. NM_197939.1
Gene Alias L13/MGC13061/Riplet
Gene Symbols RNF135
Host Species Mouse
Immunogen RNF135 (NP_922921.1, 1 a.a. ∼ 210 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 84282
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.