Learn More
ring finger protein 11, Mouse, Clone: 4G7, Abnova™
Mouse monoclonal antibody raised against a partial recombinant RNF11.
Brand: Abnova H00026994-M01.100ug
Description
The protein encoded by this gene contains a RING-H2 finger motif, which is known to be important for protein-protein interactions. The expression of this gene has been shown to be induced by mutant RET proteins (MEN2A/MEN2B). The germline mutations in RET gene are known to be responsible for the development of multiple endocrine neoplasia (MEN). [provided by RefSeq
Sequence: EEQIRIAQRIGLIQHLPKGVYDPGRDGSEKKIRECVICMMDFVYGDPIRFLPCMHIYHLDCIDDWLMRSFTCPSCMEPVDAALLSSYETNSpecifications
| ring finger protein 11 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_014372 | |
| RNF11 | |
| RNF11 (NP_055187, 65 a.a. ∼ 154 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 4G7 | |
| Mouse monoclonal antibody raised against a partial recombinant RNF11. | |
| RNF11 | |
| CGI-123/MGC51169/SID1669 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 26994 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.