Learn More
RIP Mouse, Unlabeled, Clone: 38, BD
Mouse Monoclonal Antibody
£157.25 - £284.36
Specifications
| Antigen | RIP |
|---|---|
| Clone | 38 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15805479
|
BD Biosciences
610458 |
50 μg |
£164.00 £157.25 / 1 set Save £6.75 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15815479
|
BD Biosciences
610459 |
150 μg |
£297.00 £284.36 / 1 set Save £12.64 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Binding or cross linking of the Fas antigen (also known as APO-1 and CD95) is known to elicit apoptosis in susceptible cells. Fas is a member of a family of cell surface receptors which includes tumor necrosis factor receptors (TNF-R, and TNF-R2) and nerve growth factor receptors (NGF-R), CD40, OX40, CD30, CD27, and 4-1BB. Several members of this family have been shown to regulate or induce cell death (TNF-R1 and TNF-R2). A 74kDa member of this family protein named RIP (Receptor Interacting Protein) contains an N-terminal region with homology to protein kinases and a C-terminal region containing a cytoplasmic “death domain” present in both Fas and TNF-R1. Both Fas and RIP have been shown to require this death domain to induce apoptosis and overexpression of RIP has been shown to induce cell death in transfected cells.
Immunofluorescence, Immunoprecipitation, Western Blotting
Specifications
| RIP | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG2a | |
| Affinity Purified | |
| Receptor Interacting Protein |
| 38 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Canine, Chicken, Human, Mouse, Rat | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.