missing translation for 'onlineSavingsMsg'
Learn More

RIO kinase 3 (yeast), Mouse, Clone: 3G11, Abnova™

Mouse monoclonal antibody raised against a partial recombinant RIOK3.

Brand:  Abnova H00008780-M02.100ug

Product Code. 16169235

  • £303.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This gene was identified by the similarity of its product to the Aspergillus nidulans SUDD protein, an extragenic suppressor of the heat-sensitive bimD6 mutation that fails to attach properly to the spindle microtubules at a restrictive temperature. The specific function of this gene has not yet been determined. [provided by RefSeq

Sequence: NMLWHAGKVWLIDVSQSVEPTHPHGLEFLFRDCRNVSQKGGVKEALSERELFNAVSGLNITADNEADFLAEIEALEKMNEDHVQKNGRKAASFLKDDGDPPLLYDE
Specifications

Specifications

RIO kinase 3 (yeast)
Monoclonal
Unconjugated
PBS with no preservative; pH 7.4
BC039729
RIOK3
RIOK3 (AAH39729, 411 a.a. ∼ 516 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Cell Cycle
Primary
Human
Antibody
ELISA, KnockDown, Western Blot
3G11
Mouse monoclonal antibody raised against a partial recombinant RIOK3.
RIOK3
DKFZp779L1370/SUDD
Mouse
Affinity Purified
RUO
Yes
8780
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.