missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rhox9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA523134
This item is not returnable.
View return policy
Description
Immunogen sequence homology: rat (93%), human (72%), opossum (71%).
Reproductive homeobox on X chromosome 9, also known as Reproductive Homeobox (Rhox)protein GPBOX, is a nuclear protein product of a 12 X-linked novel homeobox gene family that plays an important role during postnatal testis development. The members of this family are characterized by a conserved 60-amino-acid DNA-binding homeodomain. These transcription factors not only play an important role in various developmental processes such as DNA-dependent transcriptional regulation, body-plan specification, pattern formation, and cell fate determination, but also in understanding the development and physiology of the human reproductive system.
Specifications
| Rhox9 | |
| Polyclonal | |
| Unconjugated | |
| Rhox9 | |
| 1600026O01Rik; Gpbox; placenta specific homeobox 2; Psx2; reproductive homeobox 9; reproductive homeobox on X chromosome, 9; Rhox9; RP23-202F23.5 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| Conc. Not Determined | |
| Whole serum with 50% glycerol and 0.025% sodium azide | |
| 0 | |
| Rhox9 | |
| Within the range of amino acids 82-112 of mouse Rhox-9 protein. | |
| RUO | |
| 104384 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction