Learn More
RHOD, Mouse, Clone: 1E7, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant RHOD.
Brand: Abnova H00029984-M12.100ug
Description
Ras homolog, or Rho, proteins interact with protein kinases and may serve as targets for activated GTPase. They play a critical role in muscle differentiation. The protein encoded by this gene binds GTP and is a member of the small GTPase superfamily. It is involved in endosome dynamics and reorganization of the actin cytoskeleton, and it may coordinate membrane transport with the function of the cytoskeleton. [provided by RefSeq
Sequence: MTAAQAAGEEAPPGVRSVKVVLVGDGGCGKTSLLMVFADGAFPESYTPTVFERYMVNLQVKGKPVHLHIWDTAGQDDYDRLRPLFYPDASVLLLCFDVTSPNSFDNIFNRWYPEVNHFCKKVPIIVVGCKTDLRKDKSLVNKLRRNGLEPVTYHRGQEMARSVGAVAYLECSARLHDNVHAVFQEAAEVALSSRGRNFWRRITQGFCVVTSpecifications
| RHOD | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC001338 | |
| RHOD | |
| RHOD (AAH01338, 1 a.a. ∼ 210 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA | |
| 1E7 | |
| Mouse monoclonal antibody raised against a full-length recombinant RHOD. | |
| RHOD | |
| ARHD/RHOHP1/RHOM/Rho | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 29984 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.