missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rhinovirus VP3 Monoclonal Antibody (G47A)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA518249
This item is not returnable.
View return policy
Description
This antibody recognizes the VP3 of numerous Rhino types (6) and does not react with Enteroviruses (5), Influenza A, Influenza B, RSV, Adenovirus, Para 1, Para 2, Para 3, Chlamydia pneumoniae, Mycoplasma pneumoniae or Hepatitis A viruses. Purity is >90%.
Specifications
| Rhinovirus VP3 | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 |
| ELISA, Immunocytochemistry | |
| G47A | |
| Unconjugated | |
| rotavirus gastroenteritis | |
| Rhinovirus recombinant protein. | |
| 100 μg | |
| Primary | |
| 4°C or -20°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction